missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PCBP1 Protein

Product Code. 15922119
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15922119

Brand: enQuireBio™ QP129725ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PCBP1 was constructed and used to recombinantly synthesize the protein.

Specifications

Name PCBP1 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMDAGVTE SGLNVTLTIR LLMHGKEVGS IIGKKGESVK RIREESGARI NISEGNCPER IITLTGPTNA IFKAFAMIID KLEEDINSSM TNSTAASRPP VTLRLVVPAT QCGSLIGKGG CKIKEIREST GAQVQVAGDM LPNSTERAIT IAGVPQSVTE CVKQIC
Buffer The PCBP1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Purity or Quality Grade Greater than 85.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.