missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PCDHGC4 Protein

Product Code. 15982119
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15982119

Brand: enQuireBio™ QP129745ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PCDHGC4 was constructed and used to recombinantly synthesize the protein.

Specifications

Name PCDHGC4 Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MQIRYPVPEE SQEGTFVGNV AQDFLLDTDS LSARRLQVAG EVNQRHFRVD LDSGALLIKN PIDREALCGL SASCIVPLEF VTEGPLEMYR AEVEIVDVND HAPRFPRQQL DLEIGEAAPP GQRFPLEKAQ DADVGSNSIS SYRLSSNEHF ALDVKKRSDG SLVPELLLEK PLDREKQSDY RLVLTAVDGG NPPRSGTAEL RVSVLDVNDN APAFQQSSYR ISVLESAPAG MVLIQLNASD PDLGPSGNVT FYFSGHTPDR VRNLFSLHPT TGKLTLLGPL DFESENYYEF DVRARDGGSP AMEQHCSLRV DLLDVNDNAP YITVTSELGT LPESAEPGTV VALISVQDPD SGSNGDVSLR IPDHLPFALK SAFRNQFSLV TAGPLDREAK SSYDIMVTAS DAGNPPLSTH RTIFLNISDV NDNPPSFFQR SHEVFVPENN RPGDLLCSLA ASDPDSGLNA LISYSLLEPR NRDVSASSFI SLNPQTGAVH ATRSFDYEQT QTLQFEVQAR DRGNPPLSST VTVRLFVLDL NDNAPAVLRP RARPGSLCPQ ALPPSVGAGH LITKVTAVDL DSGYNAWVSY QLLEAPDPSL FAVSRYAGEV RTAVPIPADL PPQKLVIVVK DSGSPPLSTS VTLLVSLEED THPVVPDLRE SSAPREGESR LTLY
Buffer The PCDHGC4 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.