missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCMT1 Protein
A cDNA sequence encoding the PCMT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12976-25ug
Additional Details : Weight : 0.01000kg
Specifications
PCMT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
RGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAWK SGGASHSELI HNLRKNGIIK TDKVFEVMLA TDRSHYAKCN PYMDSPQSIG FQATISAPHM HAYALELLFD QLHEGAKALD VGSGSGILTA CFARMVGCTG KVIGIDHIKE LVDDSINNVR KDDPTLLSSG RVQLVVGDGR MGYAEEAPYD AIHVGAAAPV VPQALIDQLK PGGRLILPVG PAGGNQMLEQ YDKLQDGSIK MKPLMGVIYV PLTDKEKQWS RWK | |
Greater than 95.0% as determined by SDS-PAGE. |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PCMT1 Human solution containing 20mM Tris-HCl pH 8.0, 0.1M NaCl, and 10% glycerol. |