missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PCSK1N Protein

A cDNA sequence encoding the PCSK1N was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12981-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15992139

  • 131.00 € / 2µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

27344
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMARPVKE PRGLSAASPP LAETGAPRRF RRSVPRGEAA GAVQELARAL AHLLEAERQE RARAEAQEAE DQQARVLAQL LRVWGAPRNS DPALGLDDDP DAPAAQLARA LLRARLDPAA LAAQLVPAPV PAAALRPRPP VYDDGPAGPD AEEAGDETPD VDPELLRYLL GRILAGSADS EGVAAPRRLR RAADHDVGSE LPPEGVLGAL LRVKRLETPA PQVPARRLLP P
Greater than 85.0% as determined by SDS-PAGE.
PCSK1N Protein
Research Use Only
PCSK1N
Human
His
The PCSK1N solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers