missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human PDGF-BB Biotinylated Protein

Product Code. 18786867
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18786867

Brand: R&D Systems™ BT220010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human PDGF-BB Biotinylated Protein is derived from E. coli. The Recombinant Human PDGF-BB Biotinylated Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6FHE7
For Use With (Application) Bioactivity
Gene ID (Entrez) 5155
Molecular Weight (g/mol) Observed Molecular Wt: 12kDa, reducing conditions, Predicted Molecular Wt.: 12kDa (monomer, unlabeled)
Quantity 10 μg
Source E. coli-derived human PDGF-BB protein Ser82-Thr190
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer, biotinylated protein via amines
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Common Name PDGF-BB
Gene Symbol PDGFB
Biological Activity Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is typically 1.5-6ng/mL.
Conjugate Biotin
Species Human
Recombinant Recombinant
Expression System E. coli-derived Ser82-Thr190
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.