missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDZ and LIM domain protein 1

A DNA sequence encoding the Homo sapiens (Human) PDZ and LIM domain protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6480-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16053610

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_066272.1
PDZ and LIM domain protein 1
Research Use Only
PDLIM1
Recombinant Protein
E. coli
TTQQIDLQGP GPWGFRLVGG KDFEQPLAIS RVTPGSKAAL ANLCIGDVIT AIDGENTSNM THLEAQNRIK GCTDNLTLTV ARSEHKVWSP LVTEEGKRHP YKMNLASEPQ EVLHIGSAHN RSAMPFTASP ASSTTARVIT NQYNNPAGLY SSENISNFNN ALESKTAASG VEANSRPLDH AQPPSSLVID KESEVYKMLQ EKQELNEPPK QSTSFLVLQE ILESEEKGDP NKPSGFRSVK APVTKVAASI GNAQKLPMCD KCGTGIVGVF VKLRDRHRHP ECYVCTDCGT NLKQKGHFFV EDQIYCEKHA RERVTPPEGY EVVTVFPK
Greater than 80% as determined by SDS-PAGE.
9124
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers