missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDZ and LIM domain protein 1
A DNA sequence encoding the Homo sapiens (Human) PDZ and LIM domain protein 1, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_066272.1 |
---|---|
Gene ID (Entrez) | 9124 |
Name | PDZ and LIM domain protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043610
|
enQuireBio™
QP6480-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063610
|
enQuireBio™
QP6480-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053610
|
enQuireBio™
QP6480-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073610
|
enQuireBio™
QP6480-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083610
|
enQuireBio™
QP6480-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093610
|
enQuireBio™
QP6480-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_066272.1 | |
PDZ and LIM domain protein 1 | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9124 | |
Research Use Only | |
PDLIM1 | |
Recombinant Protein | |
E. coli | |
TTQQIDLQGP GPWGFRLVGG KDFEQPLAIS RVTPGSKAAL ANLCIGDVIT AIDGENTSNM THLEAQNRIK GCTDNLTLTV ARSEHKVWSP LVTEEGKRHP YKMNLASEPQ EVLHIGSAHN RSAMPFTASP ASSTTARVIT NQYNNPAGLY SSENISNFNN ALESKTAASG VEANSRPLDH AQPPSSLVID KESEVYKMLQ EKQELNEPPK QSTSFLVLQE ILESEEKGDP NKPSGFRSVK APVTKVAASI GNAQKLPMCD KCGTGIVGVF VKLRDRHRHP ECYVCTDCGT NLKQKGHFFV EDQIYCEKHA RERVTPPEGY EVVTVFPK | |
Greater than 80% as determined by SDS-PAGE. |