missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDZ and LIM domain protein 1
A DNA sequence encoding the Homo sapiens (Human) PDZ and LIM domain protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7666-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001107579.1 | |
PDZ and LIM domain protein 1 | |
100 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
27295 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
PDLIM3 | |
Recombinant Protein | |
E. coli | |
MPQTVILPGP APWGFRLSGG IDFNQPLVIT RITPGSKAAA ANLCPGDVIL AIDGFGTESM THADAQDRIK AAAHQLCLKI DRGETHLWSP QVSEDGKAHP FKINLESEPQ EFKPIGTAHN RRAQPFVAAA NIDDKRQVVS ASYNSPIGLY STSNIQDALH GQLRGLIPSS PQKTGTTLNT SIIFGPNLS |