missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Phenylalanine--tRNA ligase alpha subunit Protein

A DNA sequence encoding the Homo sapiens (Human) Phenylalanine--tRNA ligase alpha subunit, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8288-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013791

  • 505.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004452.1
Phenylalanine--tRNA ligase alpha subunit Protein
Research Use Only
FARSA
Recombinant Protein
E. coli
ADGQVAELLL RRLEASDGGL DSAELAAELG MEHQAVVGAV KSLQALGEVI EAELRSTKHW ELTAEGEEIA REGSHEARVF RSIPPEGLAQ SELMRLPSGK VGFSKAMSNK WIRVDKSAAD GPRVFRVVDS MEDEVQRRLQ LVRGGQAEKL GEKERSELRK RKLLAEVTLK TYWVSKGSAF STSISKQETE LSPEMISSGS WRDRPFKPYN FLAHGVLPDS GHLHPLLKVR SQFRQIFLEM GFTEMPTDNF IESSFWNFDA LFQPQQHPAR DQHDTFFLRD PAEALQLPMD YVQRVKRTHS QGGYGSQGYK YNWKLDEARK NLLRTHTTSA SARALYRLAQ KKPFTPVKYF SIDRVFRNET LDATHLAEFH QIEGVVADHG LTLGHLMGVL REFFTKLGIT QLRFKPAYNP YTEPSMEVFS YHQGLKKWVE VGNSGVFRPE MLLPMGLPEN VSVIAWGLSL ERPTMIKYGI NNIRELVGHK VNLQMVYDSP LCRLDAEPRP PPTQEAA
Greater than 90% as determined by SDS-PAGE.
2193
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers