missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phenylalanine--tRNA ligase alpha subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Phenylalanine--tRNA ligase alpha subunit, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8288-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004452.1 | |
Phenylalanine--tRNA ligase alpha subunit Protein | |
Research Use Only | |
FARSA | |
Recombinant Protein | |
E. coli | |
ADGQVAELLL RRLEASDGGL DSAELAAELG MEHQAVVGAV KSLQALGEVI EAELRSTKHW ELTAEGEEIA REGSHEARVF RSIPPEGLAQ SELMRLPSGK VGFSKAMSNK WIRVDKSAAD GPRVFRVVDS MEDEVQRRLQ LVRGGQAEKL GEKERSELRK RKLLAEVTLK TYWVSKGSAF STSISKQETE LSPEMISSGS WRDRPFKPYN FLAHGVLPDS GHLHPLLKVR SQFRQIFLEM GFTEMPTDNF IESSFWNFDA LFQPQQHPAR DQHDTFFLRD PAEALQLPMD YVQRVKRTHS QGGYGSQGYK YNWKLDEARK NLLRTHTTSA SARALYRLAQ KKPFTPVKYF SIDRVFRNET LDATHLAEFH QIEGVVADHG LTLGHLMGVL REFFTKLGIT QLRFKPAYNP YTEPSMEVFS YHQGLKKWVE VGNSGVFRPE MLLPMGLPEN VSVIAWGLSL ERPTMIKYGI NNIRELVGHK VNLQMVYDSP LCRLDAEPRP PPTQEAA | |
Greater than 90% as determined by SDS-PAGE. |
2193 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |