missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PI3 / Elafin / WFDC14 Protein

A DNA sequence encoding the Homo sapiens (Human) PI3 / Elafin / WFDC14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6493-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043700

  • 615.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002629.1
PI3 / Elafin / WFDC14 Protein
Research Use Only
PI3
Recombinant Protein
Yeast
AQEPVKGPVS TKPGSCPIIL IRCAMLNPPN RCLKDTDCPG IKKCCEGSCG MACFVPQ
Greater than 90% as determined by SDS-PAGE.
5266
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers