missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PI3K?D946GST Protein
A cDNA sequence encoding the PI3K?D946GST was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13051-1ug
Additional Details : Weight : 0.01000kg
Specifications
PI3K?D946GST Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
0.4 mg/ml solution in 25mM Tris-HCl, pH 8.0, 50mM NaCl, 0.5mM MgCl2, and 50% glycerol. |
1 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
GST | |
Greater than 95.0% as determined by SDS Page. |