missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PNPO Protein

Product Code. p-7150968
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15982459

Brand: enQuireBio™ QP130855ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PNPO was constructed and used to recombinantly synthesize the protein.

Specifications

Name PNPO Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MDPVKQFAAW FEEAVQCPDI GEANAMCLAT CTRDGKPSAR MLLLKGFGKD GFRFFTNFES RKGKELDSNP FASLVFYWEP LNRQVRVEGP VKKLPEEEAE CYFHSRPKSS QIGAVVSHQS SVIPDREYLR KKNEELEQLY QDQEVPKPKS WGGYVLYPQV MEFWQGQTNR LHDRIVFRRG LPTGDSPLGP MTHRGEEDWL YERLAP
Buffer The PNPO solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.1M NaCl and 0.1mM PMSF.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.