missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PNPT1 Protein

Product Code. p-7150969
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15912469 2 μg 2µg
15902469 10 μg 10µg
15992459 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15912469 Supplier enQuireBio™ Supplier No. QP130862ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PNPT1 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 87178
Name PNPT1 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol PNPT1
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSAVAVDLG NRKLEISSGK LARFADGSAV VQSGDTAVMV TAVSKTKPSP SQFMPLVVDY RQKAAAAGRI PTNYLRREIG TSDKEILTSR IIDRSIRPLF PAGYFYDTQV LCNLLAVDGV NEPDVLAING ASVALSLSDI PWNGPVGAVR IGIIDGEYVV NPTRKEMSSS TLNLVVAGAP KSQIVMLEAS AENILQQDFC HAIKVGVKYT QQIIQGIQQL VKETGVTKRT PQKLFTPSPE IVKYTHKLAM ERLYAVFTDY EHDKVSRDEA VNKIRLDTEE QLKEKFPEAD PYEIIESFNV VAKEVFRSIV LNEYKRCDGR DLTSLRNVSC EVDMFKTLHG SALFQRGQTQ VLCTVTFDSL ESGIKSDQVI TAINGIKDKN FMLHYEFPPY ATNEIGKVTG LNRRELGHGA LAEKALYPVI PRDFPFTIRV TSEVLESNGS SSMASACGGS LALMDSGVPI SSAVAGVAIG LVTKTDPEKG EIEDYRLLTD ILGIEDYNGD MDFKIAGTNK GITALQADIK LPGIPIKIVM EAIQQASVAK KEILQIMNKT ISKPRASRKE NGPVVETVQV PLSKRAKFVG PGGYNLKKLQ AETGVTISQV DEETFSVFAP TPSAMHEARD FITEICKDDQ EQQLEFGAVY TATITEIRDT GVMVKLYPNM TAVLLHNTQL DQRKIKHPTA LGLEVGQEIQ VKYFGRDPAD GRMRLSRKVL QSPATTVVRT LNDRSSIVMG EPISQSSSNS Q
Buffer PNPT1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Purity or Quality Grade Greater than 85.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.