missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POLL Protein

A cDNA sequence encoding the POLL was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13094-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932489

  • 181.00 € / 10µg
Estimated Shipment: 17-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

POLL Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The POLL solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 0.1M NaCl and 30% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MLMHHQKYLQ RFLGGKREKK QKEACSIPGI GKRMAEKIIE ILESGHLRKL DHISESVPVL ELFSNIWGAG TKTAQMWYQQ GFRSLEDIRS QASLTTQQAI GLKHYSDFLE RMPREEATEI EQTVQKAAQA FNSGLLCVAC GSYRRGKATC GDVDVLITHP DGRSHRGIFS RLLDSLRQEG FLTDDLVSQE ENGQQQKYLG VCRLPGPGRR HRRLDIIVVP YSEFACALLY FTGSAHFNRS MRALAKTKGM SLSEHALSTA VVRNTHGCKV GPGRVLPTPT EKDVFRLLGL PYREPAERDW
Videos
SDS
Documents
Certificates
Special Offers

Special Offers