missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human POLR1C Protein

Product Code. p-7150978
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15982489

Brand: enQuireBio™ QP130952ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the POLR1C was constructed and used to recombinantly synthesize the protein.

Specifications

Name POLR1C Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMAASQAV EEMRSRVVLG EFGVRNVHTT DFPGNYSGYD DAWDQDRFEK NFRVDVVHMD ENSLEFDMVG IDAAIANAFR RILLAEVPTM AVEKVLVYNN TSIVQDEILA HRLGLIPIHA DPRLFEYRNQ GDEEGTEIDT LQFRLQVRCT RNPHAAKDSS DPNELYVNHK VYTRHMTWIP LGNQADLFPE GTIRPVHDDI LIAQLRPGQE IDLLMHCVKG IGKDHAKFSP VATASYRLLP DITLLEPVEG EAAEELSRCF SPGVIEVQEV QGKKVARVAN PRLDTFSREI FRNEKLKKVV RLARVRDHYI FSVESTGVLP PDVLVSEAIK VLMGKCRRFL DELDAVQMD
Buffer POLR1C protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Purity or Quality Grade Greater than 80.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.