missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPA2 Protein
A cDNA sequence encoding the PPA2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13114-1mg
Additional Details : Weight : 0.01000kg
Specifications
PPA2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSALYHTEE RGQPCSQNYR LFFKNVTGHY ISPFHDIPLK VNSKEENGIP MKKARNDEYE NLFNMIVEIP RWTNAKMEIA TKEPMNPIKQ YVKDGKLRYV ANIFPYKGYI WNYGTLPQTW EDPHEKDKST NCFGDNDPID VCEIGSKILS CGEVIHVKIL GILALIDEGE TDWKLIAINA NDPEASKFHD IDDVKKFKPG YLEATLNWFR LYKVPDGKPE NQFAFNGEFK NKAFALEVIK STHQCWKALL MKKCNGGAIN CTNVQISDSP FRCTQEEARS LVESVSSSPN KESNEEEQVW HFLGK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PPA2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol, 1mM DTT. |