missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRDX4 Protein

A cDNA sequence encoding the PRDX4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10840-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945328

  • 3275.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PRDX4 Protein
Research Use Only
Specific activity: approximately 230-310 pmole/min/microg. Enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX4 and peroxide for 20 min at room temperature. Specific activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25 C for 1 minute.
Human
His
PRDX4 Human solution containing 20mM Tris-HCl pH 8 and 10% glycerol.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MWETEERPRT REEECHFYAG GQVYPGEASR VSVADHSLHL SKAKISKPAP YWEGTAVIDG EFKELKLTDY RGKYLVFFFY PLDFTFVCPT EIIAFGDRLE EFRSINTEVV ACSVDSQFTH LAWINTPRRQ GGLGPIRIPL LSDLTHQISK DYGVYLEDSG HTLRGLFIID DKGILRQITL NDLPVGRSVD ETLRLVQAFQ YTDKHGEVCP AGWKPGSETI IPDPAGKLKY FDKLN
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers