missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRDX4 Protein
A cDNA sequence encoding the PRDX4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10840-25ug
Additional Details : Weight : 0.01000kg
Specifications
PRDX4 Protein | |
Research Use Only | |
Specific activity: approximately 230-310 pmole/min/microg. Enzymatic activity was confirmed by measuring the remaining peroxide after incubation of PRDX4 and peroxide for 20 min at room temperature. Specific activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25 C for 1 minute. | |
Human | |
His | |
PRDX4 Human solution containing 20mM Tris-HCl pH 8 and 10% glycerol. |
25 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MWETEERPRT REEECHFYAG GQVYPGEASR VSVADHSLHL SKAKISKPAP YWEGTAVIDG EFKELKLTDY RGKYLVFFFY PLDFTFVCPT EIIAFGDRLE EFRSINTEVV ACSVDSQFTH LAWINTPRRQ GGLGPIRIPL LSDLTHQISK DYGVYLEDSG HTLRGLFIID DKGILRQITL NDLPVGRSVD ETLRLVQAFQ YTDKHGEVCP AGWKPGSETI IPDPAGKLKY FDKLN | |
Greater than 95% as determined by SDS-PAGE. |