missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKAA1 Protein
A cDNA sequence encoding the PRKAA1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13138-5ug
Additional Details : Weight : 0.01000kg
Specifications
PRKAA1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMRRLSSW RKMATAEKQK HDGRVKIGHY ILGDTLGVGT FGKVKVGKHE LTGHKVAVKI LNRQKIRSLD VVGKIRREIQ NLKLFRHPHI IKLYQVISTP SDIFMVMEYV SGGELFDYIC KNGRLDEKES RRLFQQILSG VDYCHRHMVV HRDLKPENVL LDAHMNAKIA DFGLSNMMSD GEFLRTSCGS PNYAAPEVIS GRLYAGPEVD IWSSGVILYA LLCGTLPFDD DHVPTLFKKI CDGIFYTPQY LNPSVISLLK HMLQVDPMKR ATIKDIREHE WF | |
Greater than 85% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRKAA1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |