missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PRND Protein

Product Code. p-7151032
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15912659 2 μg 2µg
15902659 10 μg 10µg
15992649 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15912659 Supplier enQuireBio™ Supplier No. QP131492ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PRND was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 23627
Name PRND Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol PRND
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSRGIKHRI KWNRKALPST AQITEAQVAE NRPGAFIKQG RKLDIDFGAE GNRYYEANYW QFPDGIHYNG CSEANVTKEA FVTGCINATQ AANQGEFQKP DNKLHQQVLW RLVQELCSLK HCEFWLERG
Buffer PRND protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.2M NaCl, 30% glycerol and 1mM DTT.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.