missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRNP Protein
A cDNA sequence encoding the PRNP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13150-10ug
Additional Details : Weight : 0.01000kg
Specifications
5621 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKKRPKPGGW NTGGSRYPGQ GSPGGNRYPP QGGGGWGQPH GGGWGQPHGG GWGQPHGGGW GQPHGGGWGQ GGGTHSQWNK PSKPKTNMKH MAGAAAAGAV VGGLGGYVLG SAMSRPIIHF GSDYEDRYYR ENMHRYPNQV YYRPMDEYSN QNNFVHDCVN ITIKQHTVTT TTKGENFTET DVKMMERVVE QMCITQYERE SQAYYQRGS | |
Greater than 90% as determined by SDS-PAGE. |
PRNP Protein | |
Research Use Only | |
PRNP | |
Human | |
His | |
PRNP protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1M Urea and 10% glycerol. |