missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proguanylin Protein

A cDNA sequence encoding the Proguanylin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13157-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932679

  • 203.00 € / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Proguanylin Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS VTVQDGNFSF SLESVKKLKD LQEPQEPRVG KLRNFAPIPG EPVVPILCSN PNFPEELKPL CKEPNAQEIL QRLEEIAEDP GTCEICAYAA CTGC
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Proguanylin filtered (0.4 microm) and lyophilized in 0.5 mg/ml in deionized H20.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers