missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Proteasome subunit alpha type-7 Protein
A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit alpha type-7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8360-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002783.1 | |
Proteasome subunit alpha type-7 Protein | |
Research Use Only | |
PSMA7 | |
Recombinant Protein | |
E. coli | |
RAITVFSPDG HLFQVEYAQE AVKKGSTAVG VRGRDIVVLG VEKKSVAKLQ DERTVRKICA LDDNVCMAFA GLTADARIVI NRARVECQSH RLTVEDPVTV EYITRYIASL KQRYTQSNGR RPFGISALIV GFDFDGTPRL YQTDPSGTYH AWKANAIGRG AKSVREFLEK NYTDEAIETD DLTIKLVIKA LLEVVQSGGK NIELAVMRRD QSLKILNPEE IEKYVAEI | |
Greater than 90% as determined by SDS-PAGE. |
5688 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |