missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasome subunit beta type-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8364-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034241

  • 2365.00 € / 1mg
Estimated Shipment: 28-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002785.1
Proteasome subunit beta type-2 Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
5690
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PSMB2
Recombinant Protein
E. coli
MEYLIGIQGP DYVLVASDRV AASNIVQMKD DHDKMFKMSE KILLLCVGEA GDTVQFAEYI QKNVQLYKMR NGYELSPTAA ANFTRRNLAD CLRSRTPYHV NLLLAGYDEH EGPALYYMDY LAALAKAPFA AHGYGAFLTL SILDRYYTPT ISRERAVELL RKCLEELQKR FILNLPTFSV RIIDKNGIHD LDNISFPKQG S
Videos
SDS
Documents
Certificates
Special Offers

Special Offers