missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein-arginine deiminase type-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein-arginine deiminase type-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8266-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16033651

  • 615.00 € / 100µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_031391.2
Protein-arginine deiminase type-2 Protein
Research Use Only
PADI2
Recombinant Protein
Yeast
MLRERTVRLQ YGSRVEAVYV LGTYLWTDVY SAAPAGAQTF SLKHSEHVWV EVVRDGEAEE VATNGKQRWL LSPSTTLRVT MSQASTEASS DKVTVNYYDE EGSIPIDQAG LFLTAIEISL DVDADRDGVV EKNNPKKASW TWGPEGQGAI LLVNCDRETP WLPKEDCRDE KVYSKEDLKD MSQMILRTKG PDRLPAGYEI VLYISMSDSD KVGVFYVENP FFGQRYIHIL GRRKLYHVVK YTGGSAELLF FVEGLCFPDE GFSGLVSIHV SLLEYMAQDI PLTPIFTDTV IFRIAPWIMT PNILPPVSVF VCCMKDNYLF LKEVKNLVEK TNCELKVCFQ YLNRGDRWIQ DEIEFGYIEA PHKGFPVVLD SPRDGNLKDF PVKELLGPDF GYVTREPLFE SVTSLDSFGN LEVSPPVTVN GKTYPLGRIL IGSSFPLSGG RRMTKVVRDF LKAQQVQAPV ELYSDWLTVG HVDEFMSFVP IPGTKKFLLL MASTSACYKL FREKQKDGHG EAIMFKGLGG MSSKRITINK ILSNESLVQE NLYFQRCLDW NRDILKKELG LTEQDIIDLP ALFKMDEDHR ARAFFPNMVN MIVLDKDLGI PKPFGPQVEE ECCLEMHVRG LLEPLGLECT FIDDISAYHK FLGEVHCGTN VRRKPFTFKW WHMVP
Greater than 90% as determined by SDS-PAGE.
11240
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers