missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein CYR61 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein CYR61, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5909-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958599

  • 2285.00 € / 1mg
Estimated Shipment: 17-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001545.2
Protein CYR61 Protein
Research Use Only
CYR61
Recombinant Protein
E. coli
TCPAACHCPL EAPKCAPGVG LVRDGCGCCK VCAKQLNEDC SKTQPCDHTK GLECNFGASS TALKGICRAQ SEGRPCEYNS RIYQNGESFQ PNCKHQCTCI DGAVGCIPLC PQELSLPNLG CPNPRLVKVT GQCCEEWVCD EDSIKDPMED QDGLLGKELG FDASEVELTR NNELIAVGKG SSLKRLPVFG MEPRILYNPL QGQKCIVQTT SWSQCSKTCG TGISTRVTND NPECRLVKET RICEVRPCGQ PVYSSLKKGK KCSKTKKSPE PVRFTYAGCL SVKKYRPKYC GSCVDGRCCT PQLTRTVKMR FRCEDGETFS KNVMMIQSCK CNYNCPHANE AAFPFYRLFN DIHKFRD
Greater than 90% as determined by SDS-PAGE.
3491
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers