missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein delta homolog 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein delta homolog 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5937-ma-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15928799

  • 580.00 € / 20µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001304101.1
Protein delta homolog 1 Protein
Research Use Only
DLK1
Recombinant Protein
Mammal
AECFPACNPQ NGFCEDDNVC RCQPGWQGPL CDQCVTSPGC LHGLCGEPGQ CICTDGWDGE LCDRDVRACS SAPCANNRTC VSLDDGLYEC SCAPGYSGKD CQKKDGPCVI NGSPCQHGGT CVDDEGRASH ASCLCPPGFS GNFCEIVANS CTPNPCENDG VCTDIGGDFR CRCPAGFIDK TCSRPVTNCA SSPCQNGGTC LQHTQVSYEC LCKPEFTGLT CVKKRALSPQ QVTRLPSGYG LAYRLTPGVH ELPVQQPEHR ILKVSMKELN KKTPLLTEGQ
Greater than 90% as determined by SDS-PAGE.
8788
20 μg
Not determined.
Partial (see sequence information for more details).
Human
Flag
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers