missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein-lysine N-methyltransferase N6AMT2 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein-lysine N-methyltransferase N6AMT2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7833-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031071

  • 505.00 € / 100µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001305868.1
Protein-lysine N-methyltransferase N6AMT2 Protein
Research Use Only
N6AMT2
Recombinant Protein
E. coli
SDLEDDETPQ LSAHALAALQ EFYAEQKQQI EPGEDDKYNI GIIEENWQLS QFWYSQETAL QLAQEAIAAV GEGGRIACVS APSVYQKLRE LCRENFSIYI FEYDKRFAMY GEEFIFYDYN NPLDLPERIA AHSFDIVIAD PPYLSEECLR KTSETVKYLT RGKILLCTGA IMEEQAAELL GVKMCTFVPR HTRNLANEFR CYVNYDSGLD CGI
Greater than 90% as determined by SDS-PAGE.
221143
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers