missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein phosphatase 1B Protein

A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6520-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043880

  • 518.00 € / 100µg
Estimated Shipment: 02-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001028729.1
Protein phosphatase 1B Protein
100 μg
Not determined.
Full Length of Isoform Beta-X
Human
His-SUMO
Tris-based buffer, 50% glycerol
5495
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PPM1B
Recombinant Protein
E. coli
SIVLVCFSNA PKVSDEAVKK DSELDKHLES RVEEIMEKSG EEGMPDLAHV MRILSAENIP NLPPGGGLAG KRNVIEAVYS RLNPHRESDG ASDEAEESGS QGKLVEALRQ MRINHRGNYR QLLEEMLTSY RLAKVEGEES PAEPAATATS SNSDAGNPVT MQESHTESES GLAELDSSNE DAGTKMSGEK I
Videos
SDS
Documents
Certificates
Special Offers

Special Offers