missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRRX1 Protein
A cDNA sequence encoding the PRRX1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13173-2ug
Additional Details : Weight : 0.01000kg
Specifications
PRRX1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTSSYG HVLERQPALG GRLDSPGNLD TLQAKKNFSV SHLLDLEEAG DMVAAQADEN VGEAGRSLLE SPGLTSGSDT PQQDNDQLNS EEKKKRKQRR NRTTFNSSQL QALERVFERT HYPDAFVRED LARRVNLTEA RVQVWFQNRR AKFRRNERAM LANKNASLLK SYSGDVTAVE QPIVPRPAPR PTDYLSWGTA SPYRSSSLPR CCLHEGLHNG F | |
Greater than 85.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRRX1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |