missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTGDS / L-PGDS Protein

A DNA sequence encoding the Homo sapiens (Human) PTGDS / L-PGDS, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6550-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034090

  • 2285.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000945.3
PTGDS / L-PGDS Protein
Research Use Only
PTGDS
Recombinant Protein
E. coli
APEAQVSVQP NFQQDKFLGR WFSAGLASNS SWLREKKAAL SMCKSVVAPA TDGGLNLTST FLRKNQCETR TMLLQPAGSL GSYSYRSPHW GSTYSVSVVE TDYDQYALLY SQGSKGPGED FRMATLYSRT QTPRAELKEK FTAFCKAQGF TEDTIVFLPQ TDKCMTEQ
Greater than 86% as determined by SDS-PAGE.
5730
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers