missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PTGES3 Protein

A cDNA sequence encoding the PTGES3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13201-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15952819

  • 131.00 € / 5µg
Estimated Shipment: 16-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PTGES3 Protein
Research Use Only
Recombinant Protein
E. coli
MQPASAKWYD RRDYVFIEFC VEDSKDVNVN FEKSKLTFSC LGGSDNFKHL NEIDLFHCID PNDSKHKRTD RSILCCLRKG ESGQSWPRLT KERAKLNWLS VDFNNWKDWE DDSDEDMSNF DRFSEMMNNM GGDEDVDLPE VDGADDDSQD SDDEKMPDLE
Greater than 95.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The PTGES3 protein solution contains 20mM Tris-HCl, pH 8.0, 1mM DTT and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers