missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTHrP Protein
A cDNA sequence encoding the PTHrP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13204-1mg
Additional Details : Weight : 0.01000kg
Specifications
PTHrP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
AVSEHQLLHD KGKSIQDLRR RFFLHHLIAE IHTAEIRATS EVSPNSKPSP NTKNHPVRFG SDDEGRYLTQ ETNKVETYKE QPLKTP | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
PthRp protein was lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4. |