missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PYCR1 Protein

A DNA sequence encoding the Homo sapiens (Human) PYCR1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6560-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034160

  • 505.00 € / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_008838.2
PYCR1 Protein
Research Use Only
PYCR1
Recombinant Protein
E. coli
MEQLLSSVGF CTEVEEDLID AVTGLSGSGP AYAFTALDAL ADGGVKMGLP RRLAVRLGAQ ALLGAAKMLL HSEQHPGQLK DNVSSPGGAT IHALHVLESG GFRSLLINAV EASCIRTREL QSMADQEQVS PAAIKKTILD KVKLDSPAGT ALSPSGHTKL LPRSLAPAGK D
Greater than 90% as determined by SDS-PAGE.
5831
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers