missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB24 Protein
A cDNA sequence encoding the RAB24 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13234-5ug
Additional Details : Weight : 0.01000kg
Specifications
RAB24 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMSGQRV DVKVVMLGKE YVGKTSLVER YVHDRFLVGP YQNTIGAAFV AKVMSVGDRT VTLGIWDTAG SERYEAMSRI YYRGAKAAIV CYDLTDSSSF ERAKFWVKEL RSLEEGCQIY LCGTKSDLLE EDRRRRRVDF HDVQDYADNI KAQLFETSSK TGQSVDELFQ KVAEDYVSVA AFQVMTEDKG VDLGQKPNPY FYSCCHH | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB24 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT, 0.1M NaCl, 0.1M EDTA and 30% glycerol. |