missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB3IL1 Protein
A cDNA sequence encoding the RAB3IL1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13244-2ug
Additional Details : Weight : 0.01000kg
Specifications
RAB3IL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMWSGPPQ PDQGLPPPLA AVPVPWKSTD PCQGHRESPG ALVETSAGEE AQGQEGPAAA QLDVLRLRSS SMEIREKGSE FLKEELHRAQ KELKLKDEEC ERLSKVREQL EQELEELTAS LFEEAHKMVR EANMKQAASE KQLKEARGKI DMLQAEVTAL KTLVITSTPA SPNRELHPQL LSPTKAGPRK GHSRHKSTSS TLCPAVCPAA GHTLTPDREG KEVDTILFAE FQAWRESPTL DKTCPFLERV YREDVGPCLD FTMQELSVLV RAAVEDNTLT IEPVASQTLP TVKVAEVDCS STNTCALSGL TRTCRHRIRL GDSKSHYYIS PSSRARITAV CNFFTYIRYI QQGLVRQDAE PMFWEIMRLR KEMSLAKLGF FPQEA | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RAB3IL1 protein solution (0.25 mg/ml) containing Phosphate buffer saline (pH 7.4) and 50% glycerol. |