missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAMP3 Protein
A cDNA sequence encoding the RAMP3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13260-10ug
Additional Details : Weight : 0.01000kg
Specifications
10268 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MRAGGCNETG MLERLPLCGK AFADMMGKVD VWKWCNLSEF IVYYESFTNC TEMEANVVGC YWPNPLAQGF ITGIHRQFFS NCTVDRVHLE DPPDEV | |
Greater than 90% as determined by SDS-PAGE. |
RAMP3 Protein | |
Research Use Only | |
RAMP3 | |
Human | |
His | |
RAMP3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |