missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RANKL / OPGL / TNFSF11 / CD254 Protein
Recombinant Protein
Brand: enQuireBio™ QP5325-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003692.1 | |
RANKL / OPGL / TNFSF11 / CD254 Protein | |
10 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
EKAMVDGSWL DLAKRSKLEA QPFAHLTINA TDIPSGSHKV SLSSWYHDRG WAKISNMTFS NGKLIVNQDG FYYLYANICF RHHETSGDLA TEYLQLMVYV TKTSIKIPSS HTLMKGGSTK YWSGNSEFHF YSINVGGFFK LRSGEEISIE VSNPSLLDPD QDATYFGAFK VRDID |
8600 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
TNFSF11 | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |