missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RARRES2 / Chemerin Protein
Recombinant Protein
107.00 € - 4060.00 €
Specifications
Accession Number | NP_002880.1 |
---|---|
Gene ID (Entrez) | 5919 |
Name | RARRES2 / Chemerin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15925949
|
enQuireBio™
QP5349-5UG |
5 μg |
107.00 €
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905949
|
enQuireBio™
QP5349-25UG |
25 μg |
252.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985939
|
enQuireBio™
QP5349-100UG |
100 μg |
630.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915949
|
enQuireBio™
QP5349-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995939
|
enQuireBio™
QP5349-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_002880.1 | |
RARRES2 / Chemerin Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MELTEAQRRG LQVALEEFHK HPPVQWAFQE TSVESAVDTP FPAGIFVRLE FKLQQTSCRK RDWKKPECKV RPNGRKRKCL ACIKLGSEDK VLGRLVHCPI ETQVLREAEE HQETQCLRVQ RAGEDPHSFY FPGQFAFS | |
Greater than 95% as determined by SDS-PAGE. |
5919 | |
Research Use Only | |
RARRES2 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |