missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RASSF1A Protein
A cDNA sequence encoding the RASSF1A was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | RASSF1A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933049
|
enQuireBio™
QP13273-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923049
|
enQuireBio™
QP13273-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913049
|
enQuireBio™
QP13273-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RASSF1A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RASSF1A protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMSGE PELIELRELA PAGRAGKGRT RLERANALRI ARGTACNPTR QLVPGRGHRF QPAGPATHTW CDLCGDFIWG VVRKGLQCAH CKFTCHYRCR ALVCLDCCGP RDLGWEPAVE RDTNVDEPVE WETPDLSQAE IEQKIKEYNA QINSNLFMSL NKDGSYTGFI KVQLKLVRPV SVPSSKKPPS LQDARRGPGR GTSVRRRTSF YLPKDAVKHL HVLSRTRARE VIEALLRKFL VVDDPRKFAL FERAERHGQV YLRKLLDDEQ PLRLRLLAGP SDKALSFVLK ENDSGEVNWD AFSMPELHNF LRILQREEEE HLRQILQKYS YCRQKIQEAL HACPLG | |
Greater than 85.0% as determined by SDS-PAGE. |