missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Regulator of microtubule dynamics protein 1
A DNA sequence encoding the Homo sapiens (Human) Regulator of microtubule dynamics protein 3, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001291731.1 |
---|---|
Gene ID (Entrez) | 55177 |
Name | Regulator of microtubule dynamics protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031341
|
enQuireBio™
QP7878-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16051341
|
enQuireBio™
QP7878-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16041341
|
enQuireBio™
QP7878-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061341
|
enQuireBio™
QP7878-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071341
|
enQuireBio™
QP7878-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081341
|
enQuireBio™
QP7878-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001291731.1 | |
Regulator of microtubule dynamics protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55177 | |
Research Use Only | |
RMDN3 | |
Recombinant Protein | |
E. coli | |
MSRLGALGGA RAGLGLLLGT AAGLGFLCLL YSQRWKRTQR HGRSQSLPNS LDYTQTSDPG RHVMLLRAVP GGAGDASVLP SLPREGQEKV LDRLDFVLTS LVALRREVEE LRSSLRGLAG EIVGEVRCHM EENQRVARRR RFPFVRERSD STGSSSVYFT ASSGATFTDA ESEGGYTTAN AESDNERDSD KESEDGEDEV SCETVKMGRK DSLDLEEEAA SGASSALEAG GSSGLEDVLP LLQQADELHR GDEQGKREGF QLLLNNKLVY GSRQDFLWRL ARAYSDMCEL TEEVSEKKSY ALDGKEEAEA ALEKGDESAD CHLWYAVLCG QLAEHESIQR RIQSGFSFKE HVDKAIALQP ENPMAHFLLG RWCYQVSHLS WLEKKTATAL LESPLSATVE DALQSFLKAE ELQPGFSKAG RVYISKCYRE LGKNSEARWW MKLALELPDV TKEDLAIQKD LEELEVILRD | |
Greater than 90% as determined by SDS-PAGE. |