missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Ret Fc Chimera Protein
Description
The Recombinant Human Ret Fc Chimera Protein is derived from NS0. The Recombinant Human Ret Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P07949 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 5979 |
| Molecular Weight (g/mol) | 94 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Ret protein, (N-terminus) Human Ret (Leu29-Arg635) Accession &Num; P07949 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Specific Reactivity | Ret |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?