missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ripk3 Protein
A DNA sequence encoding the Homo sapiens (Human) Ripk3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9945-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006862.2 | |
Ripk3 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
11035 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
Ripk3 | |
Recombinant Protein | |
Yeast | |
MSCVKLWPSG APAPLVSIEE LENQELVGKG GFGTVFRAQH RKWGYDVAVK IVNSKAISRE VKAMASLDNE FVLRLEGVIE KVNWDQDPKP ALVTKFMENG SLSGLLQSQC PRPWPLLCRL LKEVVLGMFY LHDQNPVLLH RDLKPSNVLL DPELHVKLAD FGLSTFQGGS QSGTGSGEPG GTLGYLAPEL FVNVNRKAST ASDVYSFGIL MWAVLAGREV ELPTEPSLVY EAVCNRQNRP SLAELPQAGP ETPGLEGLKE LMQLCWSSEP KDRPSFQECL PKTDEVFQMV ENNMNAAVST VKDFLSQLRS SNRRFSIPES GQGGTEMDGF RRTIENQHSR NDVMVSEWLN KLNLEEPPSS VPKKCPSLTK RSRAQEEQVP QAWTAGTSSD SMAQPPQTPE TSTFRNQMPS PTSTGTPSPG PRGNQGAERQ GMNWSCRTPE PNPVTGRPLV NIYNCSGVQV GDNNYLTMQQ TTALPTWGLA PSGKGRGLQH PPPVGSQEGP KDPEAWSRPQ GWYNHSGK |