missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RNA exonuclease 4 Protein

Product Code. p-7147947
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 15982758

Brand: enQuireBio™ QP10018iv10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens(Human) RNA exonuclease 4, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001266278.1
Gene ID (Entrez) 57109
Name RNA exonuclease 4 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol REXO4
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence MGKAKVPASK RAPSSPVAKP GPVKTLTRKK NKKKKRFWKS KAREVSKKPA SGPGAVVRPP KAPEDFSQNW KALQEWLLKQ KSQAPEKPLV ISQMGSKKKP KIIQQNKKET SPQVKGEEMP AGKDQEASRG SVPSGSKMDR RAPVPRTKAS GTEHNKKGTK ERTNGDIVPE RGDIEHKKRK AKEAAPAPPT EEDIWFDDVD PADIEAAIGP EAAKIARKQL GQSEGSVSLS LVKEQAFGGL TRALALDCEM VGVGPKGEES MAARVSIVNQ YGKCVYDKYV KPTEPVTDYR TAVSGIRPEN LKQGEELEVV QKEVAEMLKG RILVGHALHN DLKVLFLDHP KKKIRDTQKY KPFKSQVKSG RPSLRLLSEK ILGLQVQQAE HCSIQDAQAA MRLYVMVKKE WESMARDRRP LLTAPDHCSD DA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.