missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RQCD1 Protein

A cDNA sequence encoding the RQCD1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13361-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933329

  • 203.00 € / 20µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RQCD1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MHSLATAAPV PTTLAQVDRE KIYQWINELS SPETRENALL ELSKKRESVP DLAPMLWHSF GTIAALLQEI VNIYPSINPP TLTAHQSNRV CNALALLQCV ASHPETRSAF LAAHIPLFLY PFLHTVSKTR PFEYLRLTSL GVIGALVKTD EQEVINFLLT TEIIPLCLRI MESGSELSKT VATFILQKIL LDDTGLAYIC QTYERFSHVA MILGKMVLQL SKEPSARLLK HVVRCYLRLS DNPRAREALR QCLPDQLKDT TFAQVLKDDT TTKRWLAQLV KNLQEGQVTD PRGIPLPPQ
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The RQCD1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers