missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RTP4 Protein
A cDNA sequence encoding the RTP4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13371-20ug
Additional Details : Weight : 0.01000kg
Specifications
RTP4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMVVDFWT WEQTFQELIQ EAKPRATWTL KLDGNLQLDC LAQGWKQYQQ RAFGWFRCSS CQRSWASAQV QILCHTYWEH WTSQGQVRMR LFGQRCQKCS WSQYEMPEFS SDSTMRILSN LVQHILKKYY GNGTRKSPEM PVILEVSLEG SHDTANCEAC TLGICGQGLK SCMTKPSKSL LPHLKTGNSS PGIGAVYLAN QAKNQSAEAK EAKGSGYEKL GPSRDPD | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RTP4 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |