missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ryanodine receptor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Ryanodine receptor 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6633-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044690

  • 518.00 € / 100µg
Estimated Shipment: 28-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000531.2
Ryanodine receptor 1 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
6261
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RYR1
Recombinant Protein
E. coli
MGDAEGEDEV QFLRTDDEVV LQCSATVLKE QLKLCLAAEG FGNRLCFLEP TSNAQNVPPD LAICCFVLEQ SLSVRALQEM LANTVEAGVE SSQGGGHRTL LYGHAILLRH AHSRMYLSCL TTSRSMTDKL AFDVGLQEDA TGEACWWTMH PASKQRSEGE KVRVGDDIIL VSVSSERYLH LSTASGELQV DASFMQTLWN MNPICSRCEE GFVTGGHVLR LFHGHMDECL TISPADSDDQ RRLVYYEGGA VCTHARSLWR LEPLRISWSG SHLRWGQPLR VRHVTTGQYL ALTEDQGLVV VDASKAHTKA TSFCFRISKE KLDVAPKRDV EGMGPPEIKY GESLCFVQHV ASGLWLTYAA PDPKALRLGV LKKKAMLHQE GHMDDALSLT RCQQEESQAA RMIHSTNGLY NQFIKSLDSF SGKPRGSGPP AGTALPIEGV ILSLQDLIIY FEPPSEDLQH EEKQSKLRSL RNRQSLFQEE GMLSMVLNCI DRLNVYTTAA HFAEFAGEEA AESWKEIVNL LYELLASLIR GNRS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers