missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human SCF/c-kit Ligand Biotinylated Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human CD117/c-kit Fc Chimera (Catalog # 10961-SR ) is immobilized at 1.00 μg/mL (100 μL/well), Biotinylated Recombinant Human SCF/c-kit Ligand binds with an ED50 of 1.00-6.00 ng/mL.
Specifications
Specifications
| Accession Number | P21583.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 4254 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 20-35 kDa, under reducing conditions.; MolecularWeight-theroretical: 19 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human SCF/c-kit Ligand protein Glu26-Ala189 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | c-kit Ligand, DCUA, DFNA69, DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, FPHH, KIT ligand, Kitl, KITLG, KL-1, Mast cell growth factor, MGF, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, SHEP7, SLF, steel factor |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction