missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SCGB3A2 Protein

A cDNA sequence encoding the SCGB3A2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13429-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15913539

  • 203.00 € / 10µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SCGB3A2 Protein
Research Use Only
Recombinant Protein
E. coli
MFLINKVPLP VDKLAPLPLD NILPFMDPLK LLLKTLGISV EHLVEGLRKC VNELGPEASE AVKKLLEALS HLVGGSGGSF LINKVPLPVD KLAPLPLDNI LPFMDPLKLL LKTLGISVEH LVEGLRKCVN ELGPEASEAV KKLLEALSHL VHHHHHH
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SCGB3A2 solution at a concentration of 0.4 mg/ml in phosphate buffered saline (PBS) pH 8.0.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers